Learn More
CELLTREAT Scientific Products Glass Fiber Syringe Filter, 1.1m, 30mm, Sterile
SDP
Supplier:  CELLTREAT Scientific Products 229768
Encompass_Preferred
CELLTREAT Scientific Products 1.1¿m GF Syringe Filter. Filters are molded in the polypropylene housings, providing a reliable seal without the use of adhesives. Female luer-lock inlet and make luer slip outlet. Color coded by membrane type for easy identification. GF (Glass Fiber). Can be used as a preliminary filtering step to avoid clogging. Coarse particulate removal. Pre-filtration. Membrane diameter 30mm. RNase/DNase free and non-pyrogenic. Every case is printed with the lot number for quality traceability. Packaged sterile, individually wrapped in peel-to-open paper/plastic blister packs. 30/case.
Catalog No. 50-114-9109