Learn More
Med Vet International Dynarex insulin Syringe, 27g, 1/2" Needle, Bagged in 10S, 1Cc, 500/Case
Catalog No. 502830558
Encompass_Preferred
Dynarex Insulin Syringes are designed to deliver insulin effectively and precisely. These sterile, disposable Syringes are constructed with securely attached stainless-steel needles to help prevent needle pop-off for increased safety. The Insulin Syringes feature lubricated thin-walled, double-beveled needles for smoother insertion and improved patient comfort. Dynarex Insulin Syringes have transparent barrels with large-font gradations to allow for clear visualization.  Dynarex Insulin Syringes are high-quality hypodermic syringes designed to effectively deliver insulin. Each disposable Syringe features a sterile, stainless-steel needle with a lubricated double-beveled tip for improved patient comfort. The transparent barrel is clearly marked with large-font numbering for accurate dosing and measurement.
Catalog No. 50-283-0558 Supplier Med Vet International Supplier No. 6955
Dynarex Insulin Syringes are designed to deliver insulin effectively and precisely. These sterile, disposable Syringes are constructed with securely attached stainless-steel needles to help prevent needle pop-off for increased safety. The Insulin Syringes feature lubricated thin-walled, double-beveled needles for smoother insertion and improved patient comfort. Dynarex Insulin Syringes have transparent barrels with large-font gradations to allow for clear visualization.  Dynarex Insulin Syringes are high-quality hypodermic syringes designed to effectively deliver insulin. Each disposable Syringe features a sterile, stainless-steel needle with a lubricated double-beveled tip for improved patient comfort. The transparent barrel is clearly marked with large-font numbering for accurate dosing and measurement.