Learn More
Med Vet International EasyTouch U-100 Insulin Syringes, 1ml, 28G x 0.5in, 100/Box
Supplier:  Med Vet International 828155
Encompass_Preferred
Color-Coded to Gauge Easy-to-read information on each side of the box Needles are made with Surgical Steel Triple-Bevel Cut for a Sharper Point Thin Wall Cannulas for an Easier Flow Electro-polished to Remove Burrs for a Smoother Injection Film Coated for Maximum Comfort
Catalog No. 50-252-2511