Learn More
Med Vet International Dynarex Securesafe Safety insulin Syringe - 1 mL (New Mechanism), 29g, 1/2" Needle, 500/Case
Catalog No. 502831488
Encompass_Preferred
The Dynarex SecureSafe Insulin Syringes are high-quality hypodermic syringes designed to deliver insulin effectively and precisely. Each sterile, disposable syringe is securely attached to prevent needle pop-off and features a locking protective sheath that helps prevent needlestick injuries. The needle is coated with medical-grade lubricant and has a double-beveled tip for smoother insertion and improved patient comfort. The transparent barrel is clearly marked with bold font for accurate dose measurement.
Catalog No. 50-283-1488 Supplier Med Vet International Supplier No. 8904
The Dynarex SecureSafe Insulin Syringes are high-quality hypodermic syringes designed to deliver insulin effectively and precisely. Each sterile, disposable syringe is securely attached to prevent needle pop-off and features a locking protective sheath that helps prevent needlestick injuries. The needle is coated with medical-grade lubricant and has a double-beveled tip for smoother insertion and improved patient comfort. The transparent barrel is clearly marked with bold font for accurate dose measurement.