Learn More
Instech Pump 11 Elite inf/wd programmable dual syringe pump (704505), US cord
SDP
Catalog No. 5081063
Encompass
Syringe pump, Harvard Apparatus model 11 Elite, infuse/withdraw, programmable, dual syringe. Touch screen interface. US power cord.
Catalog No. 50-810-63 Supplier Instech Supplier No. HA1100WD
Syringe pump, Harvard Apparatus model 11 Elite, infuse/withdraw, programmable, dual syringe. Touch screen interface. US power cord.