Learn More
Instech PHD Ultra push/pull programmable syringe pump (703009), US cord
SDP
Catalog No. 5081089
Encompass
Syringe pump, Harvard Apparatus PHD Ultra, programmable push/pull, dual syringe rack. US power cord. Touch screen interface. Purchase multi rack kits separately: HA3020A (6/10), HA3021A (4X140), HA3022A (microliter).
Catalog No. 50-810-89 Supplier Instech Supplier No. HA3000P/P
Add to Cart
Add to Cart
Request a Quote
Syringe pump, Harvard Apparatus PHD Ultra, programmable push/pull, dual syringe rack. US power cord. Touch screen interface. Purchase multi rack kits separately: HA3020A (6/10), HA3021A (4X140), HA3022A (microliter).