Learn More
Med Vet International Dynarex Pre-Filled Saline Flush Syringe, 10mL, 480/Case
Catalog No. 502832300
Encompass_Preferred
Dynarex Pre-Filled Saline Flush Syringes are convenient, integrated needle-free systems designed to safely and efficiently flush compatible intravenous administration sets. These sterile, disposable syringes come pre-filled to reduce the risk of cross-contamination and minimize syringe-generated blood reflux. Clearly marked with graduations for accurate dosing, the syringes feature an easy screw-tip cap to minimize the risk of microbial contamination. Dynarex Pre-Filled Saline Flush Syringes are high-quality integrated needle-free systems designed to effectively maintain the patency of indwelling intravenous access devices. These sterile Syringes come pre-filled for time-saving convenience, and to help reduce the risk of cross-contamination and syringe-generated blood reflux.
Catalog No. 50-283-2300 Supplier Med Vet International Supplier No. 7059
Dynarex Pre-Filled Saline Flush Syringes are convenient, integrated needle-free systems designed to safely and efficiently flush compatible intravenous administration sets. These sterile, disposable syringes come pre-filled to reduce the risk of cross-contamination and minimize syringe-generated blood reflux. Clearly marked with graduations for accurate dosing, the syringes feature an easy screw-tip cap to minimize the risk of microbial contamination. Dynarex Pre-Filled Saline Flush Syringes are high-quality integrated needle-free systems designed to effectively maintain the patency of indwelling intravenous access devices. These sterile Syringes come pre-filled for time-saving convenience, and to help reduce the risk of cross-contamination and syringe-generated blood reflux.