Learn More
Trajan Scientific SYR PTFE TIP PLUNGER 50F-GT
SDP
Catalog No. 502530342
Encompass_Preferred
SGE Gastight Syringes: Fixed Needle Models, Volume: 50 uL, Needle Gauge: 25 G, Model: 25F-GT, Length Needle: 50 mm, 1.96 in., Accuracy: +/-1%, Connection Type: Fixed, Diameter Outer: 0.31 in., 8 mm, Diameter Outer Needle: 0.01 in., 0.47 mm
Catalog No. 50-253-0342 Supplier Trajan Scientific Supplier No. 004200
SGE Gastight Syringes: Fixed Needle Models, Volume: 50 uL, Needle Gauge: 25 G, Model: 25F-GT, Length Needle: 50 mm, 1.96 in., Accuracy: +/-1%, Connection Type: Fixed, Diameter Outer: 0.31 in., 8 mm, Diameter Outer Needle: 0.01 in., 0.47 mm