Learn More
Instech Syringe rack kit, 4X140 multi-rack for Harvard Apparatus PHD Ultra pumps
SDP
Catalog No. 5081100
Encompass
The HA3021 (70-3021) is a 4x140 Syringe Multi-Rack Kit for use with Harvard Apparatus PHD ULTRA series syringe pumps.
Features
    Capacity: Holds up to four 140 ml syringes.Compatibility: Designed specifically for the PHD ULTRA pump series (Standard, I/O, and Remote models).Functionality: It replaces or expands the existing syringe rack to allow for high-volume or multi-channel infusion applications.
Catalog No. 50-811-00 Supplier Instech Supplier No. HA3021
The HA3021 (70-3021) is a 4x140 Syringe Multi-Rack Kit for use with Harvard Apparatus PHD ULTRA series syringe pumps.
Features
    Capacity: Holds up to four 140 ml syringes.Compatibility: Designed specifically for the PHD ULTRA pump series (Standard, I/O, and Remote models).Functionality: It replaces or expands the existing syringe rack to allow for high-volume or multi-channel infusion applications.