Learn More
Instech Syringe rack kit, microliter rack for Harvard Apparatus PHD Ultra pumps
SDP
Supplier:  Instech HA3022A
Encompass
The HA3022A (70-3022A) is an Add-on Microdialysis Syringe Rack for immediate installation on a new Harvard Apparatus PHD ULTRA syringe pump at the time of initial purchase.
Features
    Purpose: This rack is specifically designed for sensitive applications like microdialysis and cellular injection, which require smooth and extremely low flow rates.Capacity: The rack independently holds up to four syringes.Syringe Compatibility: It is compatible with a wide range of small syringes, from 0.5 µl to 10 ml capacity.Compatibility: It is for use exclusively with the PHD ULTRA series of infusion/withdrawal syringe pumps.
Catalog No. 50-811-03