Learn More
Instech Syringe rack kit, microliter rack for Harvard Apparatus PHD Ultra pumps
SDP
Catalog No. 5081102
Encompass
The HA3022 (70-3022) is an Add-on Microdialysis Syringe Rack Kit for the Harvard Apparatus PHD ULTRA syringe pump series.
Features
    Capacity: Holds up to four microliter syringes.Syringe Range: Compatible with syringes from 0.5 µl to 10 ml.Specialty Use: Specifically designed for microdialysis applications requiring extremely low and smooth flow rates.Independent Control: Features independent thumb screws that allow for the simultaneous infusion of different-sized syringes.
Catalog No. 50-811-02 Supplier Instech Supplier No. HA3022
The HA3022 (70-3022) is an Add-on Microdialysis Syringe Rack Kit for the Harvard Apparatus PHD ULTRA syringe pump series.
Features
    Capacity: Holds up to four microliter syringes.Syringe Range: Compatible with syringes from 0.5 µl to 10 ml.Specialty Use: Specifically designed for microdialysis applications requiring extremely low and smooth flow rates.Independent Control: Features independent thumb screws that allow for the simultaneous infusion of different-sized syringes.