Learn More
Trajan Scientific SYR RHEO/VALCO VLV 100F-LC
SDP
Catalog No. 502530330
Encompass_Preferred
Trajan Syringes for Rheodyne and Valco Valves, Volume: 100 uL, Connection Type: Fixed, Length Needle: 51 mm, 2 in., Needle Gauge: 22 G, Diameter Outer Needle: 0.028 in., 0.71 mm, For Use With: For Rheodyne and Valco valves, Model: 100F-LC, Needle Point Style: Blunt, Quantity: Each
Catalog No. 50-253-0330 Supplier Trajan Scientific Supplier No. 005300
Trajan Syringes for Rheodyne and Valco Valves, Volume: 100 uL, Connection Type: Fixed, Length Needle: 51 mm, 2 in., Needle Gauge: 22 G, Diameter Outer Needle: 0.028 in., 0.71 mm, For Use With: For Rheodyne and Valco valves, Model: 100F-LC, Needle Point Style: Blunt, Quantity: Each