Learn More
Med Vet International Dynarex Securesafe Syringe w/ Safety Needle - Luer Lock, 25g, 5/8" Needle, 3 Cc, 800/Case
Catalog No. 502830936
Encompass_Preferred
The Dynarex SecureSafe Syringes with Safety Needles provide a sterile, effective way to deliver comfortable injections. All needles are color coded to represent the gauge of the needle and attach to the syringe by either luer lock or luer slip connection. The needles have a double bevel, allowing for ease of penetration while reducing the required plunger force, helping larger doses flow smoothly. The SecureSafe safety mechanism features a quick cover that allows the user to cover and protect the needle with a press and snap. The Dynarex SecureSafe Syringes with Safety Needle are designed with a safety engineered mechanism to provide a comfortable, effective way to deliver medication or draw blood, while protecting the users from accidental needlesticks.
Catalog No. 50-283-0936 Supplier Med Vet International Supplier No. 8928
The Dynarex SecureSafe Syringes with Safety Needles provide a sterile, effective way to deliver comfortable injections. All needles are color coded to represent the gauge of the needle and attach to the syringe by either luer lock or luer slip connection. The needles have a double bevel, allowing for ease of penetration while reducing the required plunger force, helping larger doses flow smoothly. The SecureSafe safety mechanism features a quick cover that allows the user to cover and protect the needle with a press and snap. The Dynarex SecureSafe Syringes with Safety Needle are designed with a safety engineered mechanism to provide a comfortable, effective way to deliver medication or draw blood, while protecting the users from accidental needlesticks.