Learn More
Med Vet International 1mL (1Cc) Luer Lock Tuberculin Syringe With Safety Needle, 25g X 5/8in, 50/Box
Supplier:  Med Vet International 27044
Encompass_Preferred
1mL (1cc) Luer Lock Tuberculin Syringe With Safety Needle, 25G X 5/8in, 50/Box Safety Needle Features One handed activation No solid surface required Simple finger stroke activation Protective arm keeps fingers behind the needle An audible "click" as well as tactile feel and a visual indicator confirm safety mechanism engagement Needle remains locked inside activated cover Fits any brand Syringe
Catalog No. 50-283-0515