Learn More
Trajan Scientific SYR UNIV 10UL SK-10FX-5C 6/PK
SDP
Catalog No. 502530403
Encompass_Preferred
Trajan SuperFlex Microsyringes, Length Needle: 50 mm, Needle Gauge: 26 G, Volume: 10 uL, Connection Type: Fixed, Diameter Outer Needle: 0.47 mm, Material: SuperfleX, Needle Point Style: Cone
Catalog No. 50-253-0403 Supplier Trajan Scientific Supplier No. 002133
Trajan SuperFlex Microsyringes, Length Needle: 50 mm, Needle Gauge: 26 G, Volume: 10 uL, Connection Type: Fixed, Diameter Outer Needle: 0.47 mm, Material: SuperfleX, Needle Point Style: Cone