Learn More
Med Vet International Easy Touch U-100 Insulin Syringe, 1/2cc 30g x 5/16in, 100/Box
Catalog No. 502092890
Encompass_Preferred
In convenient packages of 10. EasyTouch Insulin Syringes are designed and manufactured to deliver the highest level of comfort to the consumer. Our special needles are triple-bevel cut to a super sharp point, electro-polished to remove microscopic burrs, and film coated for a smoother injection.
Catalog No. 50-209-2890 Supplier Med Vet International Supplier No. 830565
In convenient packages of 10. EasyTouch Insulin Syringes are designed and manufactured to deliver the highest level of comfort to the consumer. Our special needles are triple-bevel cut to a super sharp point, electro-polished to remove microscopic burrs, and film coated for a smoother injection.