Learn More
Med Vet International Terumo Tuberculin Syringe & Needle, 1mL, Removable Needle, 25GX 5/8in, 100/BX, SS-01T2516
Catalog No. 502092895
Encompass_Preferred
Terumo SS-01T2516 1mL Syringe & Needle Combo comes with a clear barrel and easy to read graduation marks. The syringe contains a removable needle. This Terumo SS-01T2516 1mL Syringe & Needle Combo comes with a 25GX 5/8' Regular Wall needle. Regular wall needles are more rigid than thin wall needles, providing extra support and less flex due to added annula thickness. This is a tuberculin syringe. They are small syringes with fine needles used to administer medication under the skin and perform tuberculosis tests. This quantity is 100/BX
Catalog No. 50-209-2895 Supplier Med Vet International Supplier No. 01T2516
 Restricted Item
Add to Cart
Add to Cart
Terumo SS-01T2516 1mL Syringe & Needle Combo comes with a clear barrel and easy to read graduation marks. The syringe contains a removable needle. This Terumo SS-01T2516 1mL Syringe & Needle Combo comes with a 25GX 5/8' Regular Wall needle. Regular wall needles are more rigid than thin wall needles, providing extra support and less flex due to added annula thickness. This is a tuberculin syringe. They are small syringes with fine needles used to administer medication under the skin and perform tuberculosis tests. This quantity is 100/BX