Learn More
Med Vet International Easy Touch Insulin syringe, 1cc 27 x 1/2", U-100, 100/Box
Catalog No. 502092896
Encompass_Preferred
EasyTouch Insulin Syringes are designed and manufactured to deliver the highest level of comfort to the consumer. Our special needles are triple-bevel cut to a super sharp point, electro-polished to remove microscopic burrs, and film coated for a smoother injection.
Catalog No. 50-209-2896 Supplier Med Vet International Supplier No. 827155
EasyTouch Insulin Syringes are designed and manufactured to deliver the highest level of comfort to the consumer. Our special needles are triple-bevel cut to a super sharp point, electro-polished to remove microscopic burrs, and film coated for a smoother injection.