Learn More
Med Vet International AHS Syringe & Needle, 1mL, Permanent Needle, 29G X 1/2in, U-40 Insulin, 100/Box, AH10M2913-40
Catalog No. 502092904
Encompass_Preferred
AHS AH10M2913-40 1mL Syringe & Needle Combo comes with a clear barrel and easy to read graduation marks. This syringe contains a permanently attached needle. This AHS Needle is a 29G X 1/2' Regular Wall needle. Regular wall needles are more rigid than thin wall needles, providing extra support and less flex due to added annula thickness. This is an insulin syringe. It is used to inject insulin in the subcutaneous fat. This quantity is 100/BX
Catalog No. 50-209-2904 Supplier Med Vet International Supplier No. AH10M291340
AHS AH10M2913-40 1mL Syringe & Needle Combo comes with a clear barrel and easy to read graduation marks. This syringe contains a permanently attached needle. This AHS Needle is a 29G X 1/2' Regular Wall needle. Regular wall needles are more rigid than thin wall needles, providing extra support and less flex due to added annula thickness. This is an insulin syringe. It is used to inject insulin in the subcutaneous fat. This quantity is 100/BX