Learn More
Med Vet International Exel Syringe, 30-35mL, Catheter, 50/BX, 26292
Catalog No. 502559747
Encompass_Preferred
Exel International 30-35mL Syringe comes with a clear barrel and easy to read graduation marks. The catheter tip is used for flushing catheters, gastostomy tubes and other devices. This quantity is 50/BX
Catalog No. 50-255-9747 Supplier Med Vet International Supplier No. 26292
Exel International 30-35mL Syringe comes with a clear barrel and easy to read graduation marks. The catheter tip is used for flushing catheters, gastostomy tubes and other devices. This quantity is 50/BX