Learn More
Med Vet International Exel International Syringe & Needle Combo, 0.3mL, Permanent Needle, 30G x 5/16" Regular Wall, Hypodermic, 10/Pack 26014
Supplier:  Med Vet International 2601410EACH
Encompass_Preferred
Exel International .3mL Syringe comes with a clear barrel and easy to read graduation marks. This syringe contains a permanently attached needle. This Exel International 26014 Syringe & Needle Combo comes with a 30G X 5/16" Regular Wall needle. Regular wall needles are more rigid than thin wall needles, providing extra support and less flex due to added annula thickness. This hypodermic needle is a hollow needle used for withdrawing fluid like blood samples, or injecting medications. This quantity is 10/Pack The states of NY, CT, IL, NJ, NH, DE and MN restrict the sale of syringes and needles to licensed professionals and entities. By proceeding to purchase through our website, you acknowledge that you are over 18 years of age.
Catalog No. 50-209-2909
Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.