Learn More
Med Vet International Exel Syringe & Needle, .3mL, Permanent Needle, 30G X 5/16in, Insulin, 100/BX, 26014
Catalog No. 502092910
Encompass_Preferred
In convenient packages of 10. Exel International .3mL Syringe comes with a clear barrel and easy to read graduation marks. This syringe contains a permanently attached needle. This Exel International 26014 Syringe & Needle Combo comes with a 30G X 5/16' Regular Wall needle. Regular wall needles are more rigid than thin wall needles, providing extra support and less flex due to added annula thickness. This hypodermic needle is a hollow needle used for withdrawing fluid like blood samples, or injecting medications. This quantity is 100/BX
Catalog No. 50-209-2910 Supplier Med Vet International Supplier No. 26014
In convenient packages of 10. Exel International .3mL Syringe comes with a clear barrel and easy to read graduation marks. This syringe contains a permanently attached needle. This Exel International 26014 Syringe & Needle Combo comes with a 30G X 5/16' Regular Wall needle. Regular wall needles are more rigid than thin wall needles, providing extra support and less flex due to added annula thickness. This hypodermic needle is a hollow needle used for withdrawing fluid like blood samples, or injecting medications. This quantity is 100/BX