Learn More
Med Vet International Terumo Syringe, 60mL, Catheter, 25/Box, SS-60C
Catalog No. 502092929
Encompass_Preferred
Terumo SS-60C 60mL Syringe comes with a clear barrel and easy to read graduation marks. The catheter tip is used for flushing catheters, gastostomy tubes and other devices. This quantity is 25/BX
Catalog No. 50-209-2929 Supplier Med Vet International Supplier No. 60C
Terumo SS-60C 60mL Syringe comes with a clear barrel and easy to read graduation marks. The catheter tip is used for flushing catheters, gastostomy tubes and other devices. This quantity is 25/BX