Learn More
Electron Microscopy Sciences Syracuse Watch Glass 65 mm(OD) x 50 mm(ID) x 10 mm(Deep), 6/pk
SDP
Catalog No. 50949448
Encompass_Preferred
A clear watch glass which measures 65 mm(OD)x50 mm(ID)x 10 mm(Deep). The glass is grooved and has a recessed bottom which allows for stacking and prevents scratching. It is ideal for staining and specimen preparation.
Catalog No. 50-949-448 Supplier Electron Microscopy Sciences Supplier No. 7157006
A clear watch glass which measures 65 mm(OD)x50 mm(ID)x 10 mm(Deep). The glass is grooved and has a recessed bottom which allows for stacking and prevents scratching. It is ideal for staining and specimen preparation.