Learn More
Med Vet International EasyTouch Fluringe Syringe FlipLock Exchangeable Needle, 25G X 5/8in, 1mL, 50/Box
Supplier:  Med Vet International 815201
Encompass_Preferred
EasyTouch Fluringe FlipLock™ Needle Only offer an easy and simple solution in caregiver protection. After injection, just flip the protective shield over the needle until it clicks and locks in place. Hands and fingers stay away from the exposed needle. Simple, smart and cost-effective.
Catalog No. 50-255-9658