Learn More
Bio X Cell Syrian Hamster Polyclonal IgG, 5 MG
Supplier: Bio X Cell BE0087R005MG
The polyclonal Syrian hamster IgG is purified from Syrian hamster serum. It is ideal for use as a non-reactive control IgG for Syrian hamster antibodies in most in vivo and in vitro applications.
- Isotype: Syrian hamster IgG
- Recommended dilution buffer: InVivoPure pH 7.0 dilution buffer
- Unconjugated product
- Formulation: PBS, pH 7.0; contains no stabilizers or preservatives
- Endotoxin level: ≤1 EU/mg (≤0.001 EU/μg)
- Purity: ≥95%
- Sterility: 0.2 μm filtration
- Molecular weight: 150 kDa
- Storage: 4°C; do not freeze
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.