Learn More
Chemglass Life Sciences Syringe Filter, 25mm Dia, 0.45um PES Membrane, Individually Wrapped, Sterile
SDP
Supplier:  Chemglass Life Sciences CGE2005045
Encompass
PES Membrane: Low affinity for proteins and extractables with substantially faster flow rates than PVDF; suitable for pre-filtration and filtration of buffers, protein solution and culture media.
Catalog No. 50-189-7576