Learn More
Cole-Parmer Cole-Parmer HPLC Syringe Filters, Non Sterile, Nylon, 30 mm, 5.0µm; 100/pk
Supplier:  Cole-Parmer 3281614
Encompass_Preferred
These nonsterile syringe filters are tested for physical properties and UV detectable extractables. Each unit features secure, enhanced female Luer-Lok™ inlet and male luer slip outlet, and solvent-resistant low extractable polypropylene housing. Select membranes available with 1.0 µm binder-free glass prefilter for high-solids sample. All syringe filters are autoclavable.
Catalog No. NC0033955