Learn More
Medchemexpress LLC HY-N1390 100mg Medchemexpress, Syringaldehyde CAS:134-96-3 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HYN1390100MG
Encompass_Preferred
Medchemexpress, HY-N1390 100mg Syringaldehyde CAS:134-96-3 Syringaldehyde is a polyphenolic compound belonging to the group of flavonoids and is found in different plant species like Manihot esculenta and Magnolia officinalis [1] . Syringaldehyde moderately inhibits COX-2 activity with an IC 50 of 3.5 μg/mL [2] . Anti-hyperglycemic and anti-inflammatory activities [1] . Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-193-3819
Item Discontinued This item has been discontinued by the Fisher Scientific channel. Please to view product availability in your area.