Learn More
Medchemexpress LLC Syringaldehyde | 134-96-3 | 182.18 | 25 G
SDP
Supplier:  Medchemexpress LLC 25GHYN1390
Encompass_Preferred
Syringaldehyde is a polyphenolic compound found in plants such as *Manihot esculenta* and *Magnolia officinalis*. It moderately inhibits COX-2 activity with an IC50 of 3.5 μg/mL, and also exhibits anti-hyperglycemic and anti-inflammatory activities.
  • Polyphenolic compound
  • Found in various plant species
  • Moderately inhibits COX-2 activity with an IC50 of 3.5 μg/mL
  • Exhibits anti-hyperglycemic activity
  • Exhibits anti-inflammatory activity
  • Inhibits COX-2 activity in a dose-dependent manner
  • Exerts anti-hyperglycemic effect in a rat model of diabetes induced by streptozotocin
  • Has anti-inflammatory activity by inhibiting cyclo-oxygenase 2 (COX-2) in mouse macrophage cell line
Catalog No. 50-003-89569