Learn More
Medchemexpress LLC Syringaldehyde | 134-96-3 | 182.18 | 50 G

Supplier: Medchemexpress LLC HYN139050G
Syringaldehyde is a polyphenolic compound derived from plants such as Manihot esculenta and Magnolia officinalis. It serves as a laboratory chemical, moderately inhibiting COX-2 activity (IC50 of 3.5 μg/mL) and demonstrating anti-hyperglycemic and anti-inflammatory properties. This product is intended for research use only.
- Moderately inhibits COX-2 activity
- Exhibits anti-hyperglycemic activity
- Exhibits anti-inflammatory activity
- Purity of 99.95%
- Appears as a light brown to brown solid
- Molecular weight of 182.18
- Formula: C9H10O4
- Melting point: 110-113°C
- Boiling point: 192-193°C at 760 mmHg
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.