Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Syringaresinol diglucoside | 66791-77-3 | MFCD24369676 | 99.7% | 742.72 | C34H46O18 | 10 MG
SDP

Catalog No. 5000296650
Encompass_Preferred

Syringaresinol diglucoside is a natural lignan glycoside isolated from plant sources such as bamboo leaves. It is provided as a high-purity analytical standard for research use in biochemical, pharmacological, and analytical studies.

  • Natural lignan glycoside isolated from plant sources.
  • High purity (99.7%) suitable for analytical and in vitro studies.
  • Molecular formula C34H46O18; molecular weight 742.72.
  • CAS number 66791-77-3 for unambiguous identification.
  • Supplied in milligram-scale quantities intended for research use.

Catalog No. 50-002-96650 Supplier Medchemexpress LLC Supplier No. HYN195810MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Syringaresinol diglucoside is a natural lignan glycoside isolated from plant sources such as bamboo leaves. It is provided as a high-purity analytical standard for research use in biochemical, pharmacological, and analytical studies.

  • Natural lignan glycoside isolated from plant sources.
  • High purity (99.7%) suitable for analytical and in vitro studies.
  • Molecular formula C34H46O18; molecular weight 742.72.
  • CAS number 66791-77-3 for unambiguous identification.
  • Supplied in milligram-scale quantities intended for research use.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.