Learn More
Trajan Scientific SYRINGE 0.5BR-HP-0.47 0.5ML
SDP
Catalog No. 502530434
Encompass_Preferred
SGE Autosampler Syringes with 42mm Cone Tip Removable Needle, Description: Not gas tight, Volume: 0.5 uL, Diameter Outer Needle: 0.47 mm, Needle Gauge: 26 G, Length Needle: 42 mm, Connection Type: Removable, For Use With: Gas chromatography, Agilent GC Autosamplers HP7673, 7683, 7693A & 6850ALS
Catalog No. 50-253-0434 Supplier Trajan Scientific Supplier No. 000400
SGE Autosampler Syringes with 42mm Cone Tip Removable Needle, Description: Not gas tight, Volume: 0.5 uL, Diameter Outer Needle: 0.47 mm, Needle Gauge: 26 G, Length Needle: 42 mm, Connection Type: Removable, For Use With: Gas chromatography, Agilent GC Autosamplers HP7673, 7683, 7693A & 6850ALS