Learn More
Trajan Scientific SYRINGE 0.5BR-HP-.63 0.5ML
SDP
Catalog No. 502550781
Encompass_Preferred
SGE Autosampler Syringes with 42mm Cone Tip Removable Needle, Description: Not gas tight, Volume: 0.5 uL, Diameter Outer Needle: 0.63 mm, Needle Gauge: 23 G, Length Needle: 42 mm, Connection Type: Removable, For Use With: Gas chromatography, Agilent GC Autosamplers HP7673, 7683, 7693A & 6850ALS
Catalog No. 50-255-0781 Supplier Trajan Scientific Supplier No. 000410
SGE Autosampler Syringes with 42mm Cone Tip Removable Needle, Description: Not gas tight, Volume: 0.5 uL, Diameter Outer Needle: 0.63 mm, Needle Gauge: 23 G, Length Needle: 42 mm, Connection Type: Removable, For Use With: Gas chromatography, Agilent GC Autosamplers HP7673, 7683, 7693A & 6850ALS