Learn More
Trajan Scientific SYRINGE 0.5BR-PE-0.47
SDP
Catalog No. 502530431
Encompass_Preferred
Trajan PerkinElmer Syringe, Length Needle: 70 mm, Needle Gauge: 26 G, Volume: 0.5 uL, Diameter Inner Needle: 0.155 mm, Diameter Outer Needle: 0.47 mm, Needle Point Style: Beveled Cone
Catalog No. 50-253-0431 Supplier Trajan Scientific Supplier No. 000475
Trajan PerkinElmer Syringe, Length Needle: 70 mm, Needle Gauge: 26 G, Volume: 0.5 uL, Diameter Inner Needle: 0.155 mm, Diameter Outer Needle: 0.47 mm, Needle Point Style: Beveled Cone