Learn More
Chemical & Chromatography Supplies Inc Hamilton 10 µL Syringe, Fixed 23 Gauge Needle, Conical Tip, 6 Pack
SDP
Catalog No. NC1149273
Encompass
This high-precision autosampler syringe, designed by Hamilton, is ideal for gas chromatography applications. With a 10 µL capacity and a fixed 23-gauge stainless steel needle, it ensures accurate and repeatable injections while minimizing coring. Supplied in a convenient 6-pack, it is engineered for optimal performance with Agilent autosamplers, making it a preferred choice for analytical precision.
Catalog No. NC1149273 Supplier Chemical & Chromatography Supplies Inc Supplier No. 93010725
This high-precision autosampler syringe, designed by Hamilton, is ideal for gas chromatography applications. With a 10 µL capacity and a fixed 23-gauge stainless steel needle, it ensures accurate and repeatable injections while minimizing coring. Supplied in a convenient 6-pack, it is engineered for optimal performance with Agilent autosamplers, making it a preferred choice for analytical precision.