Learn More
Trajan Scientific SYRINGE 100MR-LL-GT
SDP
Catalog No. 502530289
Encompass_Preferred
SGE Gastight Syringes: Removable Luer-Lok Models, Connection Type: Removable, Diameter Outer: 1.59 in., 40.6 mm, Model: 100MR-LL-GT, Needle Gauge: No Needle Included, Volume: 100.0 mL, Accuracy: +/-1%, For Use With: Suitable for gases or liquids for maximum inertness
Catalog No. 50-253-0289 Supplier Trajan Scientific Supplier No. 009760
SGE Gastight Syringes: Removable Luer-Lok Models, Connection Type: Removable, Diameter Outer: 1.59 in., 40.6 mm, Model: 100MR-LL-GT, Needle Gauge: No Needle Included, Volume: 100.0 mL, Accuracy: +/-1%, For Use With: Suitable for gases or liquids for maximum inertness