Learn More
Trajan Scientific SYRINGE 100R 100ML REM
SDP
Catalog No. 502530335
Encompass_Preferred
SGE Series II Standard Removable Needle Syringes, Volume: 100 uL, Needle Gauge: 25 G, Diameter Outer Needle: 0.5 mm, 0.01 in., Quantity: 1/Pk., Length Needle: 50 mm, 1.96 in., Accuracy: +/-1%, Connection Type: Removable, Diameter Outer: 0.31 in., 8 mm, Includes: Standard plunger
Catalog No. 50-253-0335 Supplier Trajan Scientific Supplier No. 005050
SGE Series II Standard Removable Needle Syringes, Volume: 100 uL, Needle Gauge: 25 G, Diameter Outer Needle: 0.5 mm, 0.01 in., Quantity: 1/Pk., Length Needle: 50 mm, 1.96 in., Accuracy: +/-1%, Connection Type: Removable, Diameter Outer: 0.31 in., 8 mm, Includes: Standard plunger