Learn More
Trajan Scientific SYRINGE 100UL GT HP 23/26 ND
SDP
Catalog No. 502550770
Encompass_Preferred
SGE Autosampler Syringes with 42mm Cone Tip Fixed Needle, Description: Gas tight, Volume: 100 uL, Diameter Outer Needle: 0.63/0.47 mm, Needle Gauge: 23 to 26 G, Length Needle: 42 mm, Connection Type: Fixed, For Use With: Gas chromatography, Agilent GC Autosamplers HP7673, 7683, 7693A & 6850ALS
Catalog No. 50-255-0770 Supplier Trajan Scientific Supplier No. 005668
SGE Autosampler Syringes with 42mm Cone Tip Fixed Needle, Description: Gas tight, Volume: 100 uL, Diameter Outer Needle: 0.63/0.47 mm, Needle Gauge: 23 to 26 G, Length Needle: 42 mm, Connection Type: Fixed, For Use With: Gas chromatography, Agilent GC Autosamplers HP7673, 7683, 7693A & 6850ALS