Learn More
Trajan Scientific SYRINGE 10A-GP-GT 10ML GT
SDP
Catalog No. 502530395
Encompass_Preferred
SGE Guided Plunger Syringes, Volume: 10 uL, Connection Type: Removable, Guided PTFE Tipped Plunger Syringe, Length Needle: 50 mm, 2.75 in., Needle Gauge: 26 G, Diameter Outer Needle: 0.018 in., 0.47 mm, Model: 10R-GP-GT, Needle Point Style: Bevel, Quantity: Each
Catalog No. 50-253-0395 Supplier Trajan Scientific Supplier No. 002455
SGE Guided Plunger Syringes, Volume: 10 uL, Connection Type: Removable, Guided PTFE Tipped Plunger Syringe, Length Needle: 50 mm, 2.75 in., Needle Gauge: 26 G, Diameter Outer Needle: 0.018 in., 0.47 mm, Model: 10R-GP-GT, Needle Point Style: Bevel, Quantity: Each