Learn More
Trajan Scientific SYRINGE 10F-4.2/.63C
SDP
Catalog No. 502530378
Encompass_Preferred
SGE Microvolume Syringes, Length: 1.65 in., 42 mm, Needle Gauge: 23 G, Length Needle: 1.65 in., 42 mm, Needle Point Style: Cone, Volume: 10 uL, Connection Type: Fixed, Diameter Outer: 0.63 mm, For Use With: For making microvolume injections of 1.5uL to 8uL, Borosilicate Glass
Catalog No. 50-253-0378 Supplier Trajan Scientific Supplier No. 002839
SGE Microvolume Syringes, Length: 1.65 in., 42 mm, Needle Gauge: 23 G, Length Needle: 1.65 in., 42 mm, Needle Point Style: Cone, Volume: 10 uL, Connection Type: Fixed, Diameter Outer: 0.63 mm, For Use With: For making microvolume injections of 1.5uL to 8uL, Borosilicate Glass