Learn More
Trajan Scientific SYRINGE 10F-7 70MM LENGTH 10UL
SDP
Catalog No. 502530408
Encompass_Preferred
SGE Microvolume Syringes, Length: 2.75 in., 70 mm, Needle Gauge: 26 G, Length Needle: 2.75 in., 70 mm, Needle Point Style: Bevel, Volume: 10 uL, Connection Type: Fixed, Diameter Outer: 0.47 mm, For Use With: For making microvolume injections of 1.5uL to 8uL, Borosilicate Glass
Catalog No. 50-253-0408 Supplier Trajan Scientific Supplier No. 002003
SGE Microvolume Syringes, Length: 2.75 in., 70 mm, Needle Gauge: 26 G, Length Needle: 2.75 in., 70 mm, Needle Point Style: Bevel, Volume: 10 uL, Connection Type: Fixed, Diameter Outer: 0.47 mm, For Use With: For making microvolume injections of 1.5uL to 8uL, Borosilicate Glass