Learn More
Trajan Scientific SYRINGE 10F-CTC-.63
SDP
Supplier:  Trajan Scientific 002981
Encompass_Preferred
SGE PAL Autosampler Syringes with 50mm Fixed Cone Tip Needle, Description: Not gas tight, Volume: 10 uL, Diameter Outer Needle: 0.63 mm, Needle Gauge: 23 G, Length Needle: 50 mm, Connection Type: Fixed, For Use With: Gas chromatography, CTC, LEAP, CombiPAL, GC PAL, Length: 50 mm
Catalog No. 50-253-0359