Learn More
Trajan Scientific SYRINGE 10F-GP
SDP
Catalog No. 502530397
Encompass_Preferred
SGE Guided Plunger Syringes, Volume: 10 uL, Connection Type: Fixed, Length Needle: 50 mm, 1.96 in., Needle Gauge: 26 G, Diameter Outer Needle: 0.018 in., 0.47 mm, Model: 10F-GP, Needle Point Style: Bevel, Quantity: Each
Catalog No. 50-253-0397 Supplier Trajan Scientific Supplier No. 002400
SGE Guided Plunger Syringes, Volume: 10 uL, Connection Type: Fixed, Length Needle: 50 mm, 1.96 in., Needle Gauge: 26 G, Diameter Outer Needle: 0.018 in., 0.47 mm, Model: 10F-GP, Needle Point Style: Bevel, Quantity: Each