Learn More
Trajan Scientific SYRINGE 10F-GT-7
SDP
Catalog No. 502530401
Encompass_Preferred
SGE Gastight Syringes: Fixed Needle Models, Volume: 10 uL, Needle Gauge: 26 G, Model: 10F-GT-7, Length Needle: 70 mm, 2.75 in., Accuracy: +/-1%, Connection Type: Fixed, Diameter Outer: 0.25 in., 6.5 mm, Diameter Outer Needle: 0.01 in., 0.47 mm
Catalog No. 50-253-0401 Supplier Trajan Scientific Supplier No. 002208
SGE Gastight Syringes: Fixed Needle Models, Volume: 10 uL, Needle Gauge: 26 G, Model: 10F-GT-7, Length Needle: 70 mm, 2.75 in., Accuracy: +/-1%, Connection Type: Fixed, Diameter Outer: 0.25 in., 6.5 mm, Diameter Outer Needle: 0.01 in., 0.47 mm