Learn More
Trajan Scientific SYRINGE 10F-HP-0.47 10ML
SDP
Catalog No. 502530391
Encompass_Preferred
SGE Autosampler Syringes with 42mm Cone Tip Fixed Needle, Description: Not gas tight, Volume: 10 uL, Diameter Outer Needle: 0.47 mm, Needle Gauge: 26 G, Length Needle: 42 mm, Connection Type: Fixed, For Use With: Gas chromatography, Agilent GC Autosamplers HP7673, 7683, 7693A & 6850ALS
Catalog No. 50-253-0391 Supplier Trajan Scientific Supplier No. 002800
SGE Autosampler Syringes with 42mm Cone Tip Fixed Needle, Description: Not gas tight, Volume: 10 uL, Diameter Outer Needle: 0.47 mm, Needle Gauge: 26 G, Length Needle: 42 mm, Connection Type: Fixed, For Use With: Gas chromatography, Agilent GC Autosamplers HP7673, 7683, 7693A & 6850ALS