Learn More
Trajan Scientific SYRINGE 10F-LC 10UL
SDP
Catalog No. 502530398
Encompass_Preferred
SGE Manual Syringes For Rheodyne and Valco Valves, Connection Type: Fixed, Volume: 10 uL, Model: 10F-LC, Length Needle: 50.8 mm, 2 in., Needle Gauge: 22 G, Diameter Outer Needle: 0.028 in., 0.71 mm, For Use With: Rheodyne and Valco injection valves, Length: 2 in., Needle Point Style: LC
Catalog No. 50-253-0398 Supplier Trajan Scientific Supplier No. 002301
SGE Manual Syringes For Rheodyne and Valco Valves, Connection Type: Fixed, Volume: 10 uL, Model: 10F-LC, Length Needle: 50.8 mm, 2 in., Needle Gauge: 22 G, Diameter Outer Needle: 0.028 in., 0.71 mm, For Use With: Rheodyne and Valco injection valves, Length: 2 in., Needle Point Style: LC