Learn More
Trajan Scientific SYRINGE 10F-VA8400-5/0.47
SDP
Catalog No. 502530369
Encompass_Preferred
SGE Varian/Bruker GC Autosampler Syringes, Length Needle: 50 mm, 1.96 in., Needle Gauge: 26 G, Volume: 10 uL, Connection Type: Fixed, Diameter Outer Needle: 0.018 in., 0.47 mm, Diameter Outer: 0.47 mm, For Use With: Gas chromatography, Varian/Bruker autosampler CP8400, CP8410, CP9010, CP9050
Catalog No. 50-253-0369 Supplier Trajan Scientific Supplier No. 002950
SGE Varian/Bruker GC Autosampler Syringes, Length Needle: 50 mm, 1.96 in., Needle Gauge: 26 G, Volume: 10 uL, Connection Type: Fixed, Diameter Outer Needle: 0.018 in., 0.47 mm, Diameter Outer: 0.47 mm, For Use With: Gas chromatography, Varian/Bruker autosampler CP8400, CP8410, CP9010, CP9050