Learn More
Waters Corp Waters Corporation Syringe, 10 mL, Polypropylene
Catalog No. 50793490 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
This 10 mL polypropylene syringe is commonly used as a priming syringe for HPLC pumps as part of startup kits and for maintenance procedures.
  • 10 mL capacity
  • Constructed from polypropylene
  • Utilized for priming HPLC pumps
  • Suitable for startup and maintenance
Catalog No. 50-793-490 Supplier Waters Corp Supplier No. WAT010337
This 10 mL polypropylene syringe is commonly used as a priming syringe for HPLC pumps as part of startup kits and for maintenance procedures.
  • 10 mL capacity
  • Constructed from polypropylene
  • Utilized for priming HPLC pumps
  • Suitable for startup and maintenance