Learn More
Trajan Scientific SYRINGE 10R-HP-0.63
SDP
Catalog No. 502530387
Encompass_Preferred
SGE Autosampler Syringes with 42mm Cone Tip Removable Needle, Description: Not gas tight, Volume: 10 uL, Diameter Outer Needle: 0.63 mm, Needle Gauge: 23 G, Length Needle: 42 mm, Connection Type: Removable, Diameter Outer: 0.63 mm
Catalog No. 50-253-0387 Supplier Trajan Scientific Supplier No. 002815
SGE Autosampler Syringes with 42mm Cone Tip Removable Needle, Description: Not gas tight, Volume: 10 uL, Diameter Outer Needle: 0.63 mm, Needle Gauge: 23 G, Length Needle: 42 mm, Connection Type: Removable, Diameter Outer: 0.63 mm